Would you like to log in again?
Excellence XPR Micro-Analytical Balances, METTLER TOLEDO®
Supplier: Mettler-Toledo
30594478 30355535 30594507 30594507 30594503 30594498 30594498 30594493 30355539 30594503 30594478 30532226 30532226 30594493
76443-142EA 32010 USD InStock
76443-142 CA76443-140 CA76443-150 76443-140 76443-150 CA76443-148 76292-154 76443-148 CA76443-146 76443-146 CA76443-144 CA76443-142 76443-144 76292-156
Excellence XPR Micro-Analytical Balances, METTLER TOLEDO®
Balances and Scales
The next generation of Micro-Analytical weighing has arrived. The XPR micro-analytical balances provide ultimate accuracy and quality assurance functions that are second to none.
  • Results protocol
  • Storage of methods
  • Balance ready StatusLight
  • LevelControl: Level warning with graphical levelling guide
  • Ionizer module ready, LabX ready and calibry ready
  • Upgradable to automatic powder and / or liquid dosing
The XPR Micro-Analytical Balances, successor to the XPE Micro-Analytical Balances, offer capacity up to 52 g and boast a minimum weight of 1.4 mg (according to USP). This allows for small samples to be conveniently weighed out into larger containers or labware of different sizes - simplifying the weighing processes and avoiding the need for sample transfer. The extremely high accuracy and low minimum weight leads to considerable materialand cost savings.
Features with 7" color touchscreen (glove compatible), ergoClip for direct dosing, SmartSens for touch-less door operation, SmartTrac dosing guide automatic draft shield doors, proFACT advanced internal adjustment, MinWeigh warning function and SmartGrid weighing pan.
It has seamless process, tolerance profiles, efficient operations, adjustment and routine test history, change history function, user management and Active Temperature Control system (ATC™).
Ordering information: Optional accessories make it easy to customize your XPR micro-analytical balance to perfectly fit your process needs.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty