Would you like to log in again?
Forced Convection Cleanroom Ovens, DES and DTS Series, Yamato
Supplier: Yamato
DTS-830 DES-830
76443-186EA 29268.75 USD InStock
76443-186 76443-188
Forced Convection Cleanroom Ovens, DES and DTS Series, Yamato
Ovens Standard Ovens
These forced convection cleanroom ovens feature space saving, large volume class 100 clean oven.
  • Improved visibility and operation of control panel
  • Stable cleanliness through forced circulation with rear exhaust
  • Display power consumption, CO₂ emissions and heater manipulated variables on the control panel
  • Incorporate a maximum of 99 steps, 99 patterns program controller with repeat function
  • Triac with zero-cross heater control
Offers several options such as recorder, manual/auto damper, N₂ gas introducer with flow meter and emergency switch.
DES830 convertible to high performance filter type maintaining Class 100 at stable and fluctuating temperature up to a maximum of 200 °C.
Safety features includes self-diagnostic functions (detection for temp. sensor failure, triac shortcircuit, automatic overheating prevention, heater line disconnect, main relay contact damage), earth leakage breaker, fan motor failure, key lock function, independent overheating prevention device.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty