Would you like to log in again?
DISPENSER WIPER PETG FOR 9X9IN WIPES
Supplier: S-CURVE TECHNOLOGIES
Wiper Dispenser for 9x9 wipes - Flat top and mounting keyholes - 3/16in clear PETG material
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DISPENSER WIPER PETG FOR 9X9IN WIPES 76443-224 WDL-59-PETG Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty