Would you like to log in again?
Pathfinder™ Listeria Broth, Hardy Diagnostics
Supplier: Hardy Diagnostics
K321
76443-232PK 170.8 USD InStock
76443-232
Pathfinder™ Listeria Broth, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Pathfinder™ Listeria Broth is recommended for use with environmental samples from food processing environments or food contact surfaces as a selective and differential screening medium to detect and confirm the presence of Listeria spp. after cleaning.
  • Less steps, less time, less labor, less errors
  • Read your results as early as 30 h
  • After an additional incubation, Listeria spp. organisms will cause a fluorescent reaction
  • Leak-proof caps provide security during handling and transportation
  • Tubes have excellent impact and are shatter resistant
Eliminate false positives by using our confirmatory Listeria spp. environmental media!
Four Easy Steps!
1. Remove sampling device from container, such as EnviroMax® Environmental Sampling Swab (Cat. no. 89405-022), to swab the surface.
2. Aseptically pour all the contents of the Pathfinder™ Listeria Broth tube into the swab container.
3. Incubate at 37 °C for 30 h.
4. Read results.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty