Would you like to log in again?
Fluid D with Tween, Hardy Diagnostics
Supplier: Hardy Diagnostics
U413
76443-236PK 181.28 USD InStock
76443-236
Fluid D with Tween, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Fluid D is used by the pharmaceutical and medical device industry in procedures used to ensure sterility of injectables, non-injectables, liquid products, oils and ointments, antibiotic powders, sterile pathway devices and aerosols.
  • Bottles are tough and sturdy to reduce breakage
  • Secure tight lid ensures product integrity
  • Optically clear bottles are made of clarified polypropylene for clear viewing
  • Bottles have excellent impact and chemical resistance
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty