Would you like to log in again?
TM-235 Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Supplier: Protide Pharmaceuticals
2002 2005 2001
76443-262EA 1769.45 USD InStock
76443-262 76443-260 76443-258
TM-235 Defined Serum-Free Growth Medium, Protide Pharmaceuticals
Cell Culture Supplements and Additives
Serum replacement with similar properties to TCM™ but contains additional components for cell types that have required higher concentrations of serum (typically more than 10%). TM-235™ is defined and multi-purpose. Contains highly purified, heat-treated bovine serum albumin, heat-treated bovine transferrin and recombinant insulin.
  • Defined, serum-free
  • No hormones, antibiotics, growth factors, phenol red, Ig
  • 50X concentrate
  • Can be utilized with a wide variety of basal media
Certifications: cGMP manufactured.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty