Would you like to log in again?
pZerve DMSO Free Cryopreservation Solutions, Protide Pharmaceuticals
Supplier: Protide Pharmaceuticals
5700 5720
76443-302EA 323.62 USD InStock
76443-302 76443-300
pZerve DMSO Free Cryopreservation Solutions, Protide Pharmaceuticals
Cell Culture Supplements and Additives
A unique cryopreservation solution that does not contain DMSO, fetal bovine serum, or animal protein. pZerve™ is ready to use and does not require any dilution or further processing.
  • Does not contain DMSO
  • No fetal bovine serum or other animal proteins
  • Sterile/Ready to use
Certifications: cGMP manufactured.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty