Would you like to log in again?
Puritan® Swab, Foam Tip, Hardy Diagnostics
Supplier: Hardy Diagnostics
Puritan®
2515061PF
76446-894EA 27.92 USD InStock
76446-894
Puritan® Swab, Foam Tip, Hardy Diagnostics
Swabs Swabs for Sampling
Foam has excellent collection and release properties for cell and fluid samples. High particle collection capacity.
  • Standard plastic shaft
  • Regular tip
  • Sterile
  • Individually wrapped
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty