Would you like to log in again?
VWR PLATE 96WL PP RIMMED ROUND 1ML CS50
Supplier: VWR International
Vwr* Plate, 96 Deep well, rimmed, Sterile Material: Polypropylene, round well, DNase/RNase free, Volume: 1ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR PLATE 96WL PP RIMMED ROUND 1ML CS50 76446-948 Case of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty