Would you like to log in again?
VWR® Polystyrene Assay Plates, 96- and 384-Well
Supplier: VWR
VWR®
324032 311003 324031 208104 209104 222503 223503 301006 324022 221003 210104 205503 209004 324011 214006 215006 204512 208004 324021 204003 214003 312003 324001 301012 324012 215003 210004 324002 205512
76446-996CS 352.19 USD InStock
76446-996 76446-974 76446-976 76446-954 76446-998 76446-956 76446-978 76446-958 76447-004 76447-002 76447-000 76446-980 76446-960 76446-982 76446-962 76446-984 76221-764 76446-964 76446-986 76446-988 76447-008 76447-006 76447-012 76447-010 76446-990 76446-992 76446-970 76446-972 76446-994
VWR® Polystyrene Assay Plates, 96- and 384-Well
Microplates HTS Microplates
Manufactured from high quality crystal polystyrene. They are ideal for routine assays, mixing and storage.
  • Flat bottom ideal for microscopic applications
  • V-bottom minimising residual liquid
  • U-bottom for optimising sample/cell collection
VWR® assay plates are available in clear, white or black polystyrene, with solid or clear bottom.
The bottom of the clear bottom plates is just 0.4 mm thick and extremely flat (±0.1 mm tolerance), ensuring high measurement precision and elimination of reading errors when performing cell-based assays using fluorescent or luminescent imaging. The tissue culture-treated plates are supplied lidded and sterile, in individual bags.
The bottom of the glass bottom plates is 175 µm thick, extremely flat with a tolerance of ±15 µm. They offer a higher performance than standard polystyrene plates for fluorescence assays, luminescence detection, scintillation counting and high resolution microscopy using confocal imaging.
The solid bottom plates are suitable for absorbance, fluorescence, luminescence and scintillation applications.
*Individual clear bottoms, used when light cross over from neighbouring wells needs to be minimized.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty