Would you like to log in again?
NEBNext rRNA Depletion Kit v2 (Human, Mouse, Rat) with RNA Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs
E7405L E7405L E7405S E7405S E7405X E7405X
76447-136EA 314 USD InStock
76447-136 CA76447-136 76447-134 CA76447-138 76447-138 CA76447-134
NEBNext rRNA Depletion Kit v2 (Human, Mouse, Rat) with RNA Sample Purification Beads, New England Biolabs
Assays Nucleic Acid Assays
For added convenience, this kit contains NEBNext RNA Sample Purification Beads (Agencourt® RNAClean® XP beads from Beckman Coulter).
  • Superior depletion of rRNA from human, mouse and rat RNA
  • Depletion of cytoplasmic (5S, 5.8S, 18S, 28S, human ITS, ETS) and mitochondrial (12S and 16S) rRNA
  • Compatible with a broad range of input amounts: 10 ng - 1 µg
  • Suitable for low-quality or high-quality RNA
  • Fast workflow: 2 hours, with less than 10 minutes hands-on time
The NEBNext® rRNA Depletion Kit v2 features an improved probe design targeting all subunits of human pre-rRNA (5S, 5.8S, 18S, 28S, ITS, ETS), better coverage of mouse and rat mature rRNA, as well as improved enzyme formulations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty