Would you like to log in again?
EASY-Spray™ PepMap™ Columns
Supplier: Thermo Scientific
EASY-Spray™
ES904 ES903 ES801 ES900 ES905 ES906 ES901 ES911 ES902
76449-086EA 1483.37 USD InStock
76449-086 76449-096 76449-084 76449-094 76449-088 10044-294 76449-082 76449-092 76449-090
EASY-Spray™ PepMap™ Columns
Chromatography Columns
Nanoflow LC-MS utilizes very narrow fused silica columns and as a result, even the slightest inconsistency in column connection can result in leaks and dead volumes are a frequent source of poor data, causing unstable spray and a significant loss in performance.
Trouble-shooting connection issues can be difficult and time consuming, even for experts. Thermo Scientific EASY-Spray Columns address the aforementioned issue by incorporating high-performance nanobore columns and integrating them within a radical new design. Packed with either C18 for peptide separations or with selected wide pore phases and monoliths for the separation of intact proteins,e.g. top-down proteomics or the structural identification of biotherapeutics, the 'plug and spray' approach delivers state of the art performance in a simple to use format, making research routine.
Nano columns packed with core-enhanced Accucore C4, for top-down proteomics.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty