Would you like to log in again?
SP Bel-Art Full Coverage Face Shields with Anti-Fog, Bel-Art Products, a part of SP
Supplier: Bel-Art
Bel-Art
H24874-1000 H24874-0000
76449-252CS 152.85 USD InStock
76449-252 76419-550
SP Bel-Art Full Coverage Face Shields with Anti-Fog, Bel-Art Products, a part of SP
Face Shields
Protects against exposure to bodily fluids and infectious materials.
  • Large, wrap-around design provides coverage of entire face with unrestricted visibility
  • Anti-fog coating on interior surface prevents fogging for clear vision
  • Comfortable; suited for extended wear
  • Thick top foam guard provides distance between user and shield
  • Clear PET plastic viewing area; remove protective film prior to use
  • Disposable shield; single use only
Full coverage face shields help protect the face from potential hazards such as dust, dirt, pathogens, sprays, and fluids.
Caution: Full coverage face shields with anti-fog have not been FDA cleared or approved. The product has been authorized by FDA under an EUA for use by healthcare providers as personal protective equipment. This product is only authorized for the duration of the declaration that circumstances justifying the authorization of emergency use under Section 564(b)(1) of the Act, 21 USC 360bbb-3(b)(1) unless the authorization is terminated or revoked sooner.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty