Would you like to log in again?
VWR® Sealing Films, Non-Tacky, for qPCR, Storage and Crystallisation
Supplier: VWR
VWR®
TSS-RTQNL-25 TSSRTQNL100 TSS-RTQSNL-50
76449-260CS 59.08 USD InStock
76449-260 75805-270 89134-428
VWR® Sealing Films, Non-Tacky, for qPCR, Storage and Crystallisation
Seals Microplate Seals
50 µm polyolefin films with 50 µm inert encapsulated silicone adhesive. Especially suited for qPCR, storage and protein crystallisation applications. The encapsulated silicone adhesive is non tacky until pressed against the sealing surface, at which time adhesive is released only in sealing areas to form the strongest available heat resistant seal around each well on the plate.
  • High optical clarity, minimal to no autofluorescence
  • Chemically inert; no extractables except at extreme pH; DMSO resistant for HTS
  • Heat resistant, recommended for temperatures from −70 to +100 °C
  • Certified free from DNase, RNase, and nucleic acid
Sized to fit within the edges of raised-rim 96-well plates (76,2×133,4 mm). Two end tabs assist in positioning the film on the plate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty