Would you like to log in again?
Empore™ Solid Phase Extraction Stage Tips, CDS Analytical
Supplier: CDS Analytical
Empore®
70-2019-1003-7 70-2019-1002-5 70-2019-1006-4 70-2019-1004-9 70-2019-1005-2 70-2019-1008-8 70-2019-1007-6 70-2019-1001-3EA
76449-262CS 1452.58 USD InStock
76449-262 76449-272 76449-270 76449-266 76449-276 76449-264 76449-274 76449-268
Empore™ Solid Phase Extraction Stage Tips, CDS Analytical
Pipette Tips
The original Empore SPE membrane is now available in pre-packed StageTips and are the only solution to save time and improve reproducibility of peptide / protein desalting and fractionation.
  • Save time by avoiding manual preparation of StageTips
  • Reproducibility – Standardized packing methods to avoid tip-to-tip variability present in manual preparation in the lab
  • Compatibility – Chemically resistant to organic solvents and acidic conditions and suitable for all protein and peptide desalting / fractionation applications
  • Flexibility – Combine multiple phases into one tip or increase the number of layers for increased capacity
Empore membrane SPE StageTips are cost-effective and help users avoid manual preparation of StageTips while offering standardized packing methods to improve tip-to-tip consistency.
Particle size: 12 µm
Packaging: 96 tips per pack; 960 tips, or 10 packs, per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty