Would you like to log in again?
VWR® Aluminium Foil Seals for PCR
Supplier: VWR
VWR®
F2-RLNL-500
76449-286CS 316.29 USD InStock
76449-286
VWR® Aluminium Foil Seals for PCR
Seals Microplate Seals
Precut aluminum foil seals on a roll for PCR applications. Supplied on a roll with holder for ease of use.
  • Ideal for PCR
  • Rapidly seal microplates; faster than applying single sheeted film
  • 2 mil pierceable foil
  • Certified DNase- and RNase-free
Self-standing, non-slip dispenser is supplied for ease of use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty