Would you like to log in again?
VWR CARBOY PETG 83MM DOUBLE BAGGED ST 5L
Supplier: VWR International
Vwr* Carboy, PETG, with 83mm, Double Bagged, Gamma Sterilized, USP Class VI, FDA Grade, No Spigot, ergonomic shape maximizes bench space, wide-mouth design for easy filling and cleaning, Molded in graduation marks certified to +/-5% accuracy, offers a wide variety of top connection option, Size: 5l
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARBOY PETG 83MM DOUBLE BAGGED ST 5L 76449-410 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty