Would you like to log in again?
VWR CARBOY ASSEMBLY SINGLE USE PETG 2.5L
Supplier: VWR International
Vwr* Carboy Assembly, Single Use, Material: PETG, 83B Cap, TPE Tubing w/ Dip Tube, Gamma Sterilized, Autoclavable, USP Class VI, FDA Grade materials, Clear for high visibility of the product, High-impact strength, Ergonomic shape, Leakproof, rectangular shape saves valuable bench space, Volume: 2.5L
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CARBOY ASSEMBLY SINGLE USE PETG 2.5L 76449-432 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty