Would you like to log in again?
EZBio® Pure PFA Bottles, Foxx Life Sciences
Supplier: Foxx Life Sciences
51J-2A30-FLS 51J-2370-FLS 51J-2930-FLS 51J-2530-FLS 51J-2830-FLS 51J-2B30-FLS
76449-470EA 196.68 USD InStock
76449-470 76449-480 76449-474 76449-472 76449-478 76449-476
EZBio® Pure PFA Bottles, Foxx Life Sciences
Bottles Plastic Bottles
EZBio® Pure PFA Bottles are the ultimate storage, containment, and purity solution for valuable media. They are made of a very unique and special fluoropolymer with high thermal stability and the widest temperature range. Translucent almost to the point of being transparent, these bottles are ideally suited for contents that need final inspection.
  • High purity, USP Class VI, animal-free Perfluoroalkoxy (PFA) resin
  • Temperature range: –196 to 200 °C
  • Autoclavable
  • Double bagged
  • Easy pour, leak-proof, and drip-free design ensures safe media transfer and storage
Foxx Life Sciences EZBio® pure PFA bottles are ideally suited for downstream packaging and transfer prior to final fill and finish. With stringent laboratory requirements on cleanliness and clean manufacturing, combined with the need for excellent product quality, lot traceability and thorough, complete product validation, these bottles offer the ultimate solution for containment, storage, and shipment of highly valued products/media. Each bottle comes with a seal tested cap made from the same resin as the bottle.
All EZBio® Pure PFA bottles have a extremely high purity, and are inert to virtually every chemical used in life science applications. With nearly zero extractables, these PFA bottles have a low coefficient of friction, so no adhesion to biological materials and an inherent resistance to bioburen and biofilm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty