Would you like to log in again?
SAMPLE TUBE PS W/18IN C-FLEX 15ML PK10
Supplier: Chemglass
Sample Tube, Centrifuge, 15mL, with a pre-attached 0.2um vent filter and 18in C-Flex Inlet, Polystyrene,(Purple cap), Pre-sterilized, Single use, Transfer of biopharmaceutical products and reagents in a closed system, Gas exchange transfer
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SAMPLE TUBE PS W/18IN C-FLEX 15ML PK10 76449-640 CLS-1407-T-115 Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty