Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flowtubes for Flow Cytometry Instruments, Chemglass Life Sciences

Supplier: Chemglass

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

CLS-4380-020 CLS-4380-007 CLS-4380-005 CLS-4380-010 CLS-4380-014
76449-660CS 778.2 USD InStock
76449-660 76449-670 76449-666 76449-664 76449-658
Flowtubes for Flow Cytometry Instruments, Chemglass Life Sciences
Tubes Test Tubes
Made of crystal-clear polystyrene, these 12×75 mm FlowTubes™ are designed for fluorescence activated cell sorting/flow cytometry equipment including FACS systems from BD Biosciences.

  • Compatible with FACS (BD) instruments
  • Available with 35 µm strainer cap, standard cap, or no cap
  • Clear polystyrene
  • 5 ml capacity

The diameter and wall thickness of the tube's neck is held to very tight tolerances to ensure consistent compatibility with the automated instrument. They are available with the standard two-position culture cap, no cap at all, or with a cap that contains a 35 µm nylon mesh screen, designed to assist in cell sorting/straining applications. These tubes are primarily used to collect dissociated sample for subsequent processing in automated flow cytometry instruments.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty