Would you like to log in again?
Transport Tubes, Mailing, Screw Cap, Graduated, Sterile, Chemglass Life Sciences
Supplier: Chemglass
CLS-4740-010 CLS-4740-005
76449-672CS 251.16 USD InStock
76449-672 76449-674
Transport Tubes, Mailing, Screw Cap, Graduated, Sterile, Chemglass Life Sciences
Tubes Test Tubes
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs.
  • Leak-proof, 1.25 turn screw-cap
  • Vivid, printed graduations
  • Screw-caps attached
  • Sterile, RNase-, DNase-, pyrogen-free
Used to store, transport or freeze samples, these tubes come in both 5 ml (16×60 mm) and 10 ml (16×80 mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty