Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RACK SS YELLOW NUMBER TAG F/2IN MINIBOX
Supplier: Horizon Scientific
Rack, Material: Stainless Steel, Custom, 13 tier for 2inch Mini Boxes, 25 Cell, with yellow number tag, CUSTOMER TO PROVIDE NUMBER SEQUENCE Rack Only
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
RACK SS YELLOW NUMBER TAG F/2IN MINIBOX 76449-820 CP-13-RST Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty