Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SP Wilmad-LabGlass NMR Spinner Turbines for Bruker® Spectrometers in Assorted Colors, SP Industries

Supplier: SP Industries
STB-5-GRN STB-5-BLK STB-5-BLU STB-5 STB-5-RED STB-5-YLW
76449-892EA 299.93 USD InStock
76449-892 76449-890 76449-886 76449-896 76449-894 76449-888
SP Wilmad-LabGlass NMR Spinner Turbines for Bruker® Spectrometers in Assorted Colors, SP Industries
Stirrers Stirring Tools
High quality 5 mm NMR spinner turbines exceed Bruker® specifications.

  • Assorted colors are ideal for sample identification in multi-user facilities
  • Susceptibility matched to Bruker® originals
  • Tachometer strip includes foil protection for improved life span
  • Ambient temperature turbines allow for ±25 °C variable
  • Temperature range: −20 to 60 °C

These spinner turbines are manufactured to precision standards that meet or exceed those of the spectrometer manufacturer. This attention to detail allows the turbines to be mixed with originals during sample changer operation.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty