Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
KleenGuard™ A40 Reflex® Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
KleenGuard®
47998 47999 47997 47995 47995 47996 47999 47996 47998 47997
76449-926CS 202.9 USD InStock
CA76449-926 76449-926 CA76449-918 76449-924 CA76449-922 CA76449-924 76449-918 CA76449-920 76449-922 76449-920
KleenGuard™ A40 Reflex® Coveralls, Kimberly-Clark Professional
Disposable Garments
A40 Reflex® Coveralls are made of a microporous film and laminate fabric which provides a better liquid barrier than the leading competitor.
  • Coveralls feature a respirator fit hood, zipper front, and elastic wrists and ankles
  • Breathable, microporous film laminated non-woven fabric provides barrier protection against solid particles, dust, and liquid splash
  • Reflex® design delivers enhanced comfort with fewer rip-outs
  • 3.8X better liquid barrier than leading competitor
  • 30% stronger fabric with up to 8X less lint than leading competitor
KleenGuard™ Personal Protective Equipment (PPE) enables rather than inhibits, allowing for maximum productivity and safety for staff and employees. KleenGuard™ A40 Reflex® coveralls with hood are a great addition when offering comfortable protective clothing is a must. Each garment is equipped with a respirator fit hood, zipper front, and elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates, the KleenGuard™ A40 material provides a better liquid barrier and a better particulate barrier than leading competitors.
These coveralls with hood pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids and have Reflex® sizing for a more comfortable fit and less rip outs. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing, and much more.
Certifications: Passes NFPA 99 for antistatic materials and ASTM F1670 testing for penetration of blood and bodily fluids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty