Would you like to log in again?
DuPont™ Tyvek® Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
DuPont™
BWLT2A20 BWLT2A72 BWLT2A30 BWLT2A48 BWLT2A60 BWLT2A36 BWLT2A38 BWLT2A54 BWLT2A120 BWLT2A56 BWLT2A90 BWLT2A32 BWLT2A64 BWLT2A34 BWLT2A46 BWLT2A42 BWLT2A24 BWLT2A144 BWLT2A26
76468-902CS 2053.38 USD InStock
76468-902 76450-290 76450-292 76450-284 76450-294 76450-286 76450-296 76450-310 76450-288 76450-298 76450-312 76450-300 76450-314 76450-302 76450-316 76450-304 76450-318 76450-306 76450-308
DuPont™ Tyvek® Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.
After sterilization and after the outer Tyvek® bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed.
Remaining in place over the bowl during setup, their use minimizes microbial or particulate contamination risks.
Delivery information: Certificates of conformance sent with each shipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty