Would you like to log in again?
BHD Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BWLNWB24 BWLNWB48 BWLNWB30 BWLNWB36
76450-320CS 5710.71 USD InStock
76450-320 76450-322 76450-324 76450-328
BHD Autoclavable Stopper Bowl Covers, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization.
After sterilization and after the outer bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed.
Remaining in place over the bowl during setup, their use minimizes microbial or particulate contamination risks.
Delivery information: Certificates of conformance sent with each shipment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty