Would you like to log in again?
BHD Stopper Bowl Covers, 2-Ply, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
BWL2NWB36 BWL2NWB48
76450-330CS 4283.01 USD InStock
76450-330 76450-326
BHD Stopper Bowl Covers, 2-Ply, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave in place during the stopper bowl installation to prevent microbial and particulate contamination and leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • 2-Ply for enhanced strength and resistance to punctures
  • Individually poly bagged
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
2-Ply BHD Stopper bowl covers provide a convenient, time-saving method of wrapping stopper bowls prior to steam sterilization.
After sterilization and after the outer bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed.
Remaining in place over the bowl during setup, their use minimizes microbial or particulate contamination risks.
Delivery information: Certificates of conformance sent with each shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty