Would you like to log in again?
Mini-Muff Closures, Polypropylene, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
MFNWHD16 MFNWHD07 MFNWHD08 MFNWHD09 MFNWHD04 MFNWHD05 MFNWHD06 MFNWHD12 MFNWHD18
76450-394CS 3979.02 USD InStock
76450-394 76450-382 76450-384 76450-386 76450-378 76450-388 76450-390 76450-392 76450-380
Mini-Muff Closures, Polypropylene, Keystone Cleanroom Products
Covers
Keep cleaned non-sterile production hoses protected from external sources of contamination.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Tear and puncture resistant
  • Conveniently packaged-100 per sealed poly bag
These polypropylene muffs are a great choice in non-sterile pharmaceutical manufacturing operations. Can be used as a clean equipment cover to protect process hoses.
Poly bagged in packs of 100 to help ensure cleanliness up until point of use.
Delivery information: Certificates of conformance sent with each shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty