Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DuPont™ Tyvek® Mini-Muff Closures with Skirt, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

DuPont™
MFT2A18SK02 MFT2A09SK02 MFT2A07SK02 MFT2A05K02 MFT2A12SK02 MFT2A18SK02TL
76450-396CS 8158.13 USD InStock
76450-396 76450-398 76450-400 76450-402 76450-404 76450-406
DuPont™ Tyvek® Mini-Muff Closures with Skirt, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.

  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Conveniently packaged-100 per sealed poly bag

Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs with a skirt extension provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings with the addition of a 2" long skirt extension. The skirt extension provides a safe area to remove the muff without indvertently touching the critical surfaces with the fingers.

Delivery information: Certificates of conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty