Would you like to log in again?
DuPont™ Tyvek® Mini-Muff, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
DuPont™
MFT2A04 MFT2A03 MFT2A08 MFT2A18 MFT2A09 MFT2A07 MFT2A05OT MFT2A16 MFT2A06BT MFT2A12 MFT2A05
76450-420CS 3017 USD InStock
76450-420 76450-430 76450-422 76450-432 76450-424 76450-434 76450-426 76450-436 76450-428 76450-438 76450-418
DuPont™ Tyvek® Mini-Muff, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Conveniently packaged-100 per sealed poly bag
Save time and standardize the activities in equipment preparation for sterilization. Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes and hoses. These closures are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce time consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.
Delivery information: Certificates of conformance sent with each shipment.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty