Would you like to log in again?
Tyvek® Stopper Bags, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
SBMGTVKHS0610 SBMGTVKHS1212 SBMGTVKHS1414 SBMGTVKHS1616 SBMGTVKHS0306 SBMGTVKHS0409 SBMGTVKHS1518 SBMGTVKHS0304 SBMGTVKHS0422 SBMGTVKHS1035 SBMGTVKHS1318 SBMGTVKHS0630 SBMGTVKHS2216 SBMGTVKHS0508 SBMGTVKHS2418 SBMGTVKHS1824 SBMGTVKHS1219 SBMGTVKHS1920 SBMGTVKHS2440 SBMGTVKHS2436 SBMGTVKHS1215 SBMGTVKHS0812 SBMGTVKHS1632 SBMGTVKHS2026 SBMGTVKHS1238 SBMGTVKHS2848 SBMGTVKHS2022
76450-480CS 4432.1 USD InStock
76450-480 76450-460 76450-482 76450-440 76450-484 76450-462 76450-442 76450-486 76450-464 76450-444 76450-466 76450-488 76450-468 76450-446 76450-448 76450-490 76450-470 76450-492 76450-450 76450-472 76450-452 76450-474 76450-476 76450-454 76450-456 76450-478 76450-458
Tyvek® Stopper Bags, Keystone Cleanroom Products
Bags Sterilization Pouches
1073B Tyvek® / HDPE autoclave bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps. Also a good choice for heavy equipment parts where visibility into the bag is required.
  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • Semi-transparent HDPE film side
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong, double welded factory seals
1073B Tyvek® side permits steam penetration during sterilization. These are a good choice for heavy, or pharmaceutical change parts with sharp corners or where visibility into the bag is required after sterilization.
Delivery information: Supplied with Certificates of Conformance.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty