1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges.
- Manufactured and packaged in a controlled cleanroom environment
- Steam sterilization indicator - autoclavable at 121 to 125 °C
- Tear and puncture resistant
- Hydrophobic and lint-free
- 100% Medical Grade 1073B Tyvek® permits fast steam penetration
- Excellent impact resistance with exceptional microbial barrier
- Consistently strong factory seals
100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.
With consistently strong factory seals, these are a good choice for heavy, or pharmaceutical change parts with sharp corners.
All Tyvek construction allows for a greater number of bags per sterilization load; increasing efficiencies and cost reduction.
Delivery information: Certificates of Conformance sent with each shipment.