Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyvek® 1073B Autoclave Bags with Steam Indicator, Heat Seal, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tyvek®
COVMGTVK36X60 COVMGTVK14X22 COVMGTVK18X30 COVMGTVKUB8X9 COVMGTVK40X48 COVMGTVK12X24 COVMGTVK32X34 COVMGTVK14X42 COVMGTVK24X24 COVMGTVKUB1X14 COVMGTVK18X24 COVMGTVK15X18 COVMGTVK42X46 COVMGTVK18X68 COVMGTVK20X32 COVMGTVK26X28 COVMGTVK18X40 COVMGTVK12X18 COVMGTVK48X64 COVMGTVK12X12 COVMGTVK30X32 COVMGTVK38X48 COVMGTVK25X37 COVMGTVKUB6X16 COVMGTVK16X36 COVMGTVKUB8X30 COVMGTVK48X28 COVMGTVK12X16 COVMGTVK16X32 COVMGTVKUB3X36 COVMGTVK24X44 COVMGTVKUB6X34 COVMGTVKUB6X8 COVMGTVK30X22 COVMGTVK12X32 COVMGTVKUB6X22 COVMGTVK16X44 COVMGTVK48X48 COVMGTVK10X25 COVMGTVK20X20 COVMGTVK10X20 COVMGTVK38X40 COVMGTVK16X24 COVMGTVK36X48 COVMGTVK18X26 COVMGTVK10X12 COVMGTVKUB3X6 COVMGTVK12X34 COVMGTVKUB8X12 COVMGTVKUB8X14 COVMGTVK18X22 COVMGTVK30X30 COVMGTVK36X42 COVMGTVKUB5X25 COVMGTVK34X40 COVMGTVK36X36 COVMGTVK26X30 COVMGTVK14X28 COVMGTVK34X34
76450-618CS 821.04 USD InStock
76450-618 76450-580 76450-582 76450-584 76450-540 76450-620 76450-586 76450-542 76450-622 76450-588 76450-544 76450-624 76450-546 76450-626 76450-548 76450-628 76450-608 76450-570 76450-572 76450-530 76450-574 76450-576 76450-532 76450-578 76450-534 76450-610 76450-536 76450-612 76450-538 76450-614 76450-616 76450-560 76450-562 76450-564 76450-520 76450-600 76450-566 76450-522 76450-602 76450-524 76450-568 76450-604 76450-526 76450-606 76450-528 76450-590 76450-592 76450-550 76450-594 76450-552 76450-596 76450-554 76450-598 76450-630 76450-556 76450-514 76450-558 76450-516 76450-518
Tyvek® 1073B Autoclave Bags with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
1073B Tyvek® are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps as well as heavy equipment parts with sharp edges.

  • Manufactured and packaged in a controlled cleanroom environment
  • Steam sterilization indicator - autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic and lint-free
  • 100% Medical Grade 1073B Tyvek® permits fast steam penetration
  • Excellent impact resistance with exceptional microbial barrier
  • Consistently strong factory seals

100% Medical Grade 1073B Tyvek® Autoclave Bags are ideal for steam sterilization of pharmaceutical components such as vial and syringe stoppers and caps.

With consistently strong factory seals, these are a good choice for heavy, or pharmaceutical change parts with sharp corners.

All Tyvek construction allows for a greater number of bags per sterilization load; increasing efficiencies and cost reduction.

Delivery information: Certificates of Conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty