Would you like to log in again?
DuPont™ Tyvek® Medical Grade 1073B Autoclave Wrappers, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
MGTVKSHEET2854 MGTVKSHEET2020 MGTVKSHEET0406 MGTVKSHEET1010 MGTVKSHEET0404 MGTVKSHEET0203 MGTVKSHEET1515 MGTVKSHEET6060 MGTVKSHEET0536 MGTVKSHEET0808 MGTVKSHEET0911 MGTYSHEET60140 MGTVKSHEET0810 MGTVKSHEET0306 MGTVKSHEET0836 MGTVKSHEET1818 MGTVKSHEET1212 MGTVKSHEET5454 MGTVKSHEET0606 MGTVKSHEET0707 MGTVKSHEET1836 MGTVKSHEET2430 MGTVKSHEET3636 MGTVKSHEET0518 MGTVKSHEET4040 MGSHEET7.5X7.5 MGTVKSHEET1860 MGTVKSHEET2424 MGTVKSHEET1036 MGTVKSHEET0909 MGTVKSHEET4080 MGTVKSHEET0303 MGTVKSHEET1616 MGTVKSHEET3030
76450-816CS 731.34 USD InStock
76450-816 76450-818 76450-780 76450-760 76450-782 76450-762 76450-820 76450-786 76450-764 76450-822 76450-800 76450-788 76450-824 76450-802 76450-768 76450-804 76450-806 76450-808 76450-790 76450-770 76450-794 76450-772 76450-752 76450-774 76450-796 76450-776 76450-798 76450-754 76450-756 76450-778 76450-810 76450-758 76450-812 76450-814
DuPont™ Tyvek® Medical Grade 1073B Autoclave Wrappers, Keystone Cleanroom Products
Sterilization Wraps
Medical grade 1073B Tyvek® wrappers improve the microbial barrier and cleanliness of your sterilization wrap.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
Medical grade 1073B Tyvek® wrappers are an improvement over cellulose or SMS based wrapping materials for steam sterilization. Eliminate the wasted time and expense of cutting larger square sheets of sterilization wrap to your desired sizes. Clean cut squares and rectangles are delivered in labeled, sealed poly bags to ensure cleanliness.
Delivery information: Certificates of Conformance sent with each shipment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty