Would you like to log in again?
Tyvek® 1073B / PE Steam Sterilization Chevron Peel Pouches, with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
BBMGTVKHS2435 BBMGTVKHS1012 BBMGTVKHS0415 BBMGTVKHS0308 BBMGTVKHS0208 BBMGTVKHS1214 BBMGTVKHS0814 BBMGTVKHS2418 BBMGTVKHS1620 BBMGTVKHS0510 BBMGTVKHS0620 BBMGTVKHS0625 BBMGTVKHS1011 BBMGTVKHS1516 BBMGTVKHS0612 BBMGTVKHS1015 BBMGTVKHS1424 BBMGTVKHS0807 BBTMGTVK12-30 BBMGTVKHS1624 BBMGTVKHS0206 BBMGTVKHS1218 BBMGTVKHS1030 BBMGTVKHS1020 BBMGTVKHS1618 BBMGTVKHS0815 BBMGTVKHS1217 BBTMGTVK2448 BBMGTVKHS03508 BBMGTVKHS1630 BBMGTVKHS0610 BBMGTVKHS1212 BBTMGTVK12-18 BBMGTVKHS0713 BBMGTVKHS0608 BBMGTVKHS2424 BBMGTVKHS0818 BBMGTVKHS1635 BBMGTVKHS1520 BBTMGTVK1840 BBMGTVKHS1617 BBMGTVKHS1220 BBMGTVKHS0628 BBMGTVKHS0710 BBTMGTVK2442 BBMGTVKHS1224 BBMGTVKHS0834 BBMGTVKHS1632 BBMGTVKHS0635 BBMGTVKHS0810 BBTMGTVK2440 BBMGTVKHS2430 BBMGTVKHS1822 BBMGTVKHS0306 BBMGTVKHS1420 BBMGTVKHS0422 BBMGTVKHS0632 BBMGTVKHS1626 BBMGTVKHS2426 BBMGTVKHS1616 BBMGTVKHS02095 BBMGTVKHS1816
77335-298CS 1767.36 USD InStock
77335-298 77335-330 77335-332 77335-334 77335-336 77335-290 77335-292 77335-294 77335-296 76450-860 76450-862 76637-342 77335-328 76450-864 76450-866 76450-868 76450-826 77335-320 77335-286 77335-322 77335-288 77335-324 77335-326 77335-284 76450-850 77335-316 76450-852 77335-318 76450-854 76450-856 76450-858 76450-838 77335-352 77335-310 77335-354 77335-312 77335-356 77335-358 77335-314 77335-350 76450-840 76450-842 77335-306 76450-844 77335-308 76450-846 76450-848 76450-828 77335-342 77335-300 77335-344 77335-302 77335-346 76637-344 77335-348 77335-304 77335-340 76450-830 77335-338 76450-832 76450-834 76450-836
Tyvek® 1073B / PE Steam Sterilization Chevron Peel Pouches, with Steam Indicator, Heat Seal, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly
  • Chevron peel
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Heat seals closed
  • Integrated steam sterilization indicator
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Delivery information: Certificates of Conformance sent with each shipment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty