Would you like to log in again?
Tyvek® 1073B Self-Sealing / PE Steam Sterilization Chevron Peel Pouches with Steam Indicator, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
BBMGTVKSS1617 BBMGTVKSS1822 BBMGTVKSS0810 BBMGTVKSS1020 BBMGTVKSS1626 BBMGTVKSS03508 BBMGTVKSS1635X BBMGTVKSS1214 BBMGTVKSS0308 BBMGTVKSS02095 BBMGTVKSS0632 BBMGTVKSS2430 BBMGTVKSS0713 BBMGTVKSS1635 BBMGTVKSS0612 BBMGTVKSS1520 BBMGTVKSS0415 BBMGTVKSS0635 BBMGTVKSS0206 BBMGTVKSS0306 BBMGTVKSS2426 BBMGTVKSS0608 BBMGTVKSS0510 BBMGTVKSS0834 BBMGTVKSS1217 BBMGTVKSS2435 BBMGTVKSS0422 BBMGTVKSS0818 BBMGTVKSS1012 BBMGTVKSS1632 BBMGTVKSS0815 BBMGTVKSS1424 BBMGTVKSS0628
76450-916CS 3245.52 USD InStock
76450-916 76450-918 76450-880 76450-882 76450-884 76450-886 76450-888 76450-920 76450-922 76450-900 76450-924 76450-902 76450-926 76450-904 76450-928 76450-906 76450-908 76450-890 76450-892 76450-870 76450-872 76450-894 76450-930 76450-896 76450-874 76450-910 76450-932 76450-898 76450-876 76450-912 76450-934 76450-878 76450-914
Tyvek® 1073B Self-Sealing / PE Steam Sterilization Chevron Peel Pouches with Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the prep time, performance, microbial barrier and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Easy, fast and reliable self-sealing closure system
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly, chevron peel, hydrophobic
  • Tear and puncture resistant, lint-free
  • Integrated steam sterilization indicator
A significant upgrade over paper based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper based autoclave pouches routinely do. Save valuable time in equipment preparation with the easy to use self-sealing closure system. No need for the use and validation of a heat sealer.
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterization compatible materials. These pouches are also compatible with VHP decontamination cycles commonly used with Isolator technology. Will not absorb or adsorp water.
Delivery information: Certificates of Conformance sent with each shipment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty