Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Isolator Bags, Self-Sealing Tyvek® 1073B/ PE Steam Sterilization Chevron Peel Pouch with Steam Indicator and Hang Hole, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
Tyvek®
BBMGTVKSS1217H BBMGTVKSS1424H BBMGTVKSS0209H BBMGTVKSS1012H BBMGTVKSS1617H BBMGTVKSS0815H BBMGTVKSS0628H BBMGTVKSS0834H BBMGTVKSS0612H BBMGTVKSS2435H BBMGTVKSS1626H BBMGTVKSS0635H BBMGTVKSS1020H BBMGTVKSS1822H BBMGTVKSS0510H BBMGTVKSS0818H BBMGTVKSS1520H BBMGTVKSS03508H BBMGTVKSS0415H BBMGTVKSS2426H BBMGTVKSS0308H BBMGTVKSS0306H BBMGTVKSS0713H BBMGTVKSS1214H BBMGTVKSS0422H BBMGTVKSS2430H BBMGTVKSS0608H BBMGTVKSS0632H BBMGTVKSS1632H BBMGTVKSS0810H BBMGTVKSS0206H BBMGTVKSS1635H
76450-938CS 1604.7 USD InStock
76450-938 76450-980 76450-960 76450-982 76450-984 76450-940 76450-962 76450-986 76450-942 76450-964 76450-966 76450-944 76450-988 76450-968 76450-946 76450-948 76450-990 76450-970 76450-992 76450-972 76450-950 76450-994 76450-974 76450-996 76450-952 76450-954 76450-998 76450-976 76450-978 76450-956 76450-958 76450-936
Isolator Bags, Self-Sealing Tyvek® 1073B/ PE Steam Sterilization Chevron Peel Pouch with Steam Indicator and Hang Hole, Keystone Cleanroom Products
Bags Sterilization Pouches
Improve the prep time, performance, microbial barrier, and cleanliness of your autoclave peel pouches.
  • Manufactured and packaged in a controlled cleanroom environment
  • Integrated Hang Hole is ideal for use with Isolator VHP decontamination cycles
  • Easy, fast, and reliable self-sealing closure system
  • Autoclavable at 121 to 125 °C
  • Peels open cleanly and lint-free
  • Chevron peel and hydrophobic
  • Tear and puncture resistant
  • Integrated Steam Sterilization Indicator
A significant upgrade over paper-based steam sterilization pouches. Integrated Hang-Hole for use in Isolators. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do. Save valuable time in equipment preparation with the easy-to-use self-sealing closure system.
No need for the use and validation of a heat sealer. The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb H₂O₂.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty