Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyvek® 1422A Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tyvek®
TUBT2A08 TUBT2A12 TUBT2A28 TUBT2A04 TUBT2A40 TUBT2A06 TUBT2A16 TUBT2A02 TUBT2A24 TUBT2A10 TUBT2A03 TUBT2A05 TUBT2A30 TUBT2A15
76451-018CS 2332.22 USD InStock
76451-018 76451-028 76451-016 76451-038 76451-026 76451-036 76451-014 76451-024 76451-034 76451-022 76451-032 76451-020 76451-030 76451-040
Tyvek® 1422A Steam Sterilization Tubing, Continuous Form, Keystone Cleanroom Products
Bags Sterilization Pouches
Continuous Form tubing made from 1422A Tyvek® provides the user with tremendous sizing flexibility when wrapping pharmaceutical equipment for steam sterilization.

  • Manufactured and packaged in a controlled cleanroom environment
  • On a plastic core (no corrugated material)
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free
  • Consistently strong factory seals
  • Exceptional microbial barrier

This steam sterilization tubing is a great choice when there is a wide range of equipment parts that need to be wrapped for sterilization.

Simply unroll the tubing to the desired length, slide the equipment into the tube and then heat seal each end closed.

The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.

Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficiency and throughput.

Delivery information: Certificates of Conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty