Would you like to log in again?
Tyvek® 1422A Autoclave bags with Drawstring and Steam Indicator, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
Tyvek®
BGT2A4850D BGT2A2424D BGT2A3642D BGT2A0616D BGT2A2832D BGT2A0710D BGT2A1030D BGT2A1543D BGT2A2440D BGT2A1832D BGT2A1824D BGT2A1224D BGT2A2646D BGT2A0815D BGT2A1632D BGT2A0425D BGT2A1018D BGT2A4244D BGT2A1848D BGT2A3240D BGT2A1212D BGT2A1644D BGT2A3248D BGT2A1414D BGT2A3648D BGT2A2030D BGT2A2040D BGT2A3628D BGT2A3434D BGT2A1242D BGT2A1232D BGT2A3672D BGT2A6250D BGT2A3664D BGT2A2048D BGT2A1836D BGT2A2046D BGT2A1024D BGT2A0418D BGT2A4450D BGT2A2828D BGT2A1220D BGT2A1616D BGT2A1636D BGT2A0511D BGT2A1870D BGT2A0606D BGT2A1230D BGT2A4060D BGT2A2032D BGT2A2024D BGT2A1240D BGT2A1624D BGT2A1214D BGT2A1840D BGT2A0609D BGT2A2020D BGT2A0812D BGT2A1820D BGT2A3636D BGT2A1638D BGT2A3646D BGT2A0426D BGT2A1216D BGT2A1228D BGT2A2634D BGT2A1440D BGT2A2448D BGT2A3838D BGT2A1420D BGT2A0713D BGT2A0428D BGT2A0508D BGT2A3030D BGT2A1630D BGT2A0707D BGT2A2648D BGT2A1854D BGT2A4848D BGT2A1436D BGT2A1072D BGT2A1236D BGT2A1218D BGT2A1830D BGT2A3232D BGT2A0636D BGT2A2036D BGT2A1620D BGT2A5028D BGT2A1422D BGT2A0822D BGT2A1648D BGT2A1064D BGT2A0610D BGT2A1012D BGT2A1424D BGT2A1438D BGT2A1822D BGT2A1020D BGT2A0709D BGT2A1222D BGT2A0615D BGT2A0608D BGT2A1448D BGT2A3028D BGT2A1426D BGT2A0505D18 BGT2A0506D BGT2A2044D BGT2A0510D BGT2A4040D BGT2A1526D BGT2A2436D BGT2A0509D BGT2A3436D BGT2A0811D BGT2A1846D BGT2A4864D BGT2A09512D
76451-308CS 4723.76 USD InStock
76451-308 76451-306 76451-304 76451-302 76451-148 76451-268 76451-146 76451-300 76451-266 76451-144 76451-264 76451-142 76451-262 76451-140 76451-260 76451-138 76451-258 76451-136 76451-256 76451-134 76451-254 76451-132 76451-252 76451-130 76451-250 76451-160 76451-280 76451-208 76451-328 76451-206 76451-326 76451-204 76451-324 76451-202 76451-168 76451-322 76451-288 76451-200 76451-320 76451-166 76451-286 76451-164 76451-284 76451-162 76451-282 76451-270 76451-318 76451-316 76451-314 76451-158 76451-312 76451-278 76451-310 76451-156 76918-242 76451-276 76451-154 76451-274 76451-152 76451-272 76451-150 76451-182 76451-180 76451-228 76451-226 76451-224 76451-222 76451-342 76451-188 76451-220 76451-186 76451-340 76451-184 76451-292 76468-904 76451-170 76451-290 76468-908 76468-906 76451-218 76451-338 76451-216 76451-336 76451-214 76451-334 76451-212 76451-332 76451-178 76451-298 76451-210 76451-330 76451-176 76451-296 76451-174 76451-294 76451-172 76451-128 76451-248 76451-126 76451-246 76451-124 76451-244 76451-122 76451-242 76451-120 76451-240 76451-192 76451-190 76451-118 76451-238 76451-116 76451-236 76451-114 76451-234 76451-232 76451-198 76451-230 76451-196 76451-194
Tyvek® 1422A Autoclave bags with Drawstring and Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Tyvek® 1422A autoclave bags with drawstrings can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Tear and puncture resistant
  • Fast and consistent for equipment preparation area
  • Steam Indicator
  • Hydrophobic
  • Lint-free
  • Strong factory seals
  • Exceptional microbial barrier
Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these drawstring bags will also save valuable time in the equipment prep area.
The drapeable nature of the Tyvek® 1422A material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
This creates secondary level of protection during aseptic presentation of equipment also eliminates the use of autoclave tape and heat sealers. Manufactured and packaged in a controlled cleanroom environment.
Delivery information: Certificates of Conformance sent with each shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty