Tyvek® 1422A autoclave bags with drawstrings can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
- Tear and puncture resistant
- Fast and consistent for equipment preparation area
- Steam Indicator
- Hydrophobic
- Lint-free
- Strong factory seals
- Exceptional microbial barrier
Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these drawstring bags will also save valuable time in the equipment prep area.
The drapeable nature of the Tyvek® 1422A material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.
This creates secondary level of protection during aseptic presentation of equipment also eliminates the use of autoclave tape and heat sealers. Manufactured and packaged in a controlled cleanroom environment.
Delivery information: Certificates of Conformance sent with each shipment.