Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tyvek® 1422A Autoclave Bags with Steam Indicator, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tyvek®
BGT2A1030 BGT2A0516 BGT2A0710 BGT2A0808 BGT2A3642 BGT2A0824 BGT2A2818 BGT2A2864 BGT2A1830 BGT2A0448 BGT2A2840 BGT2A1418 BGT2A4872 BGT2A1836 BGT2A2436 BGT2A1010 BGT2A09512 BGT2A0416 BGT2A1240 BGT2A0638 BGT2A2430 BGT2A0610 BGT2A2026 BGT2A2472 BGT2A2448 BGT2A3060 BGT2A0430 BGT2A1234 BGT2A1218 BGT2A1420 BGT2A2844 BGT2A1624 BGT2A4040 BGT2A0406 BGT2A1216 BGT2A1534 BGT2A4860 BGT2A0522 BGT2A0203 BGT2A4416 BGT2A1638 BGT2A606 BGT2A2040 BGT2A0105 BGT2A1620 BGT2A0310 BGT2A2222 BGT2A1424 BGT2A3016 BGT2A1543 BGT2A0820 BGT2A0309 BGT2A2440 BGT2A3434 BGT2A1232 BGT2A5054 BGT2A506 BGT2A0662 BGT2A0872 BGT2A0404 BGT2A2044 BGT2A2325 BGT2A3040 BGT2A2226 BGT2A1062 BGT2A3036 BGT2A2424 BGT2A0236 BGT2A2225 BGT2A0412 BGT2A2462 BGT2A1426 BGT2A2030 BGT2A1020 BGT2A1848 BGT2A0250 BGT2A1212 BGT2A1818 BGT2A0612 BGT2A3636 BGT2A0856 BGT2A1252 BGT2A1604 BGT2A3246 BGT2A3020 BGT2A1530 BGT2A1416 BGT2A0205 BGT2A2460 BGT2A0822 BGT2A0809 BGT2A3412 BGT2A1526 BGT2A2020 BGT2A4848 BGT2A1222 BGT2A2838 BGT2A2628 BGT2A1224 BGT2A0812 BGT2A2834 BGT2A1015 BGT2A1824 BGT2A1422 BGT2A5050 BGT2A2024 BGT2A0108 BGT2A0124 BGT2A3030 BGT2A3648 BGT2A1236 BGT2A4836 BGT2A0.7508 BGT2A0.7505 BGT2A2028 BGT2A0409 BGT2A1248 BGT2A1632 BGT2A0208 BGT2A4864 BGT2A1246 BGT2A2442 BGT2A0106 BGT2A0608 BGT2A0306 BGT2A1612 BGT2A1024 BGT2A1855 BGT2A3660 BGT2A0816 BGT2A0526 BGT2A2224 BGT2A4242
76451-380CS 2651.65 USD InStock
76451-380 76468-898 76469-860 76469-862 76451-428 76469-856 76451-426 76469-858 76469-852 76451-424 76451-422 76469-854 76451-388 76451-420 76451-386 76451-384 76451-382 76469-870 76469-872 76451-408 76451-418 76469-868 76469-900 76451-416 76469-902 76451-414 76469-864 76469-866 76451-412 76451-378 76451-410 76451-376 76469-908 76451-374 76451-372 76469-904 76469-906 76451-370 76468-878 76468-876 76469-840 76469-834 76469-836 76451-448 76451-446 76469-830 76451-444 76469-832 76451-442 76468-884 76468-882 76451-440 76469-838 76468-880 76451-390 76468-888 76468-886 76469-850 76469-846 76469-848 76451-438 76451-436 76469-842 76451-434 76469-844 76468-896 76451-432 76451-398 76468-894 76451-430 76451-396 76468-892 76451-394 76468-890 76451-392 76451-348 76451-468 76469-896 76451-346 76469-898 76451-466 76451-344 76451-464 76451-462 76451-460 76469-824 76469-826 76451-458 76469-820 76469-822 76451-456 76451-454 76468-874 76468-872 76451-452 76468-870 76451-450 76469-828 76469-882 76469-884 76469-880 76469-912 76451-406 76469-878 76469-914 76451-404 76451-402 76469-874 76451-368 76469-910 76451-400 76469-876 76451-366 76451-364 76469-916 76451-362 76451-360 76469-918 76469-892 76469-894 76469-890 76469-922 76469-886 76451-358 76469-888 76469-920 76451-356 76451-354 76451-474 76451-352 76451-472 76451-350 76451-470
Tyvek® 1422A Autoclave Bags with Steam Indicator, Keystone Cleanroom Products
Bags Sterilization Pouches
Tyvek® 1422A autoclave bags with steam indicators can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.

  • Tear and puncture resistant
  • Designed to be heat sealed closed
  • Superior to paper based autoclave pouches and wraps
  • Fast and consistent for equipment preparation area
  • Steam indicator and hydrophobic
  • Lint-free; strong factory seals
  • Exceptional microbial barrier

Standardize and simplify the way equipment is wrapped for steam sterilization. The use of these bags will also save valuable time in the equipment prep area.

The drapeable nature of the 1422A Tyvek® material is ideal for wrapping equipment parts where one may need to 'feel through' the bag; especially when lifting, or handling the wrapped part.

Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.

Manufactured and packaged in a controlled cleanroom environment.

Delivery information: Supplied with Certificate of Conformance.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty