Would you like to log in again?
BHD Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Tyvek®
BNWBG2435D2P BNWBG2032D2P BNWT3654D22P BNWBG09.512D2P BNWBG01644D2P BNWBG0606D2P BNWBG1830D2P BNWBG2436D2P BNWBG1854D2P BNWBG1212D2P BNWBG2024D2P BNWBG3648D2P BNWBG1218D2P BNWBG4040D2P BNWBG2424D2P BNWBG1624D2P BNWBG0642D2P
76451-508CS 6570.47 USD InStock
76451-508 76451-518 76451-506 76451-516 76451-504 76451-514 76451-524 76451-502 76451-512 76451-522 76451-500 76451-510 76451-498 76451-520 76451-496 76451-494 76451-492
BHD Autoclave Bags with Tyvek® Drawstring Closure, Blue, Keystone Cleanroom Products
Bags Sterilization Pouches
BHD Autoclave bags with Tyvek® Drawstrings can be used as either the inner, or outer bag for the steam sterilization of pharmaceutical change parts.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with BioShield paper
  • Eliminates need to cut material and the use of autoclave tape
Standardize and simplify the way equipment is wrapped for steam sterilization.
The use of these 2 ply drawstring bags will also save valuable time in the equipment prep area. The 2-ply BHD material resists ripping and punctures.
Since 100% of the surface area allows for steam penetration and drying, each load can be configured for maximum efficieny and throughput.
The drawstring bags can be aseptically opened without generating particles like cutting other types of bags open can.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty