Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DuPont™ Tyvek® 1422A Elasticized Covers for Steam Sterilization, Rectangular Shaped, Keystone Cleanroom Products

Supplier: Keystone Cleanroom Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Tyvek®
T2ASHEET2440E T2ASHEET3024E T2ASHEET0324E T2SHET2838E2D2 T2ASHEET4233E T2ASHEET3434E T2ASHEET1818E T2ASHEET4080E T2ASHEET4320E T2ASHEET2626E T2ASHEET2816E T2ASHEET2424E T2ASHEET3014E
76451-608CS 2528.26 USD InStock
76451-608 76451-618 76451-606 76451-628 76451-616 76451-604 76451-626 76451-614 76451-624 76451-612 76451-622 76451-610 76451-620
DuPont™ Tyvek® 1422A Elasticized Covers for Steam Sterilization, Rectangular Shaped, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave the covers in place during installation to prevent microbial and particulate contamination. Leave in place while the remaining fill line set-up activities are performed.

  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Simplifies the conformance to wrapping SOP's
  • Steam Sterilization Indicator - Autoclavable at 121 to 125 °C
  • Tear and puncture resistant
  • Hydrophobic
  • Lint-free

Elasticized Tyvek® Covers provide a convenient, time-saving method of wrapping difficult to wrap product contact change parts prior to steam sterilization. These covers allow immediate steam penetration and work best with modern pulse vacuum autoclaves for sufficient drying.

After sterilization and after the outer Tyvek® bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed.

Remaining in place over the equipment parts during setup, their use minimizes microbial or particulate contamination risks.

Certifications: Certificates of conformance sent with each shipment.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty