Would you like to log in again?
BHD Elasticized Covers for Steam Sterilization, Rectangular Shaped with Steam Indicator, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
BHDSHEET2440E BHDSHT2838E2D2
76451-794CS 3615.47 USD InStock
76451-794 76451-792
BHD Elasticized Covers for Steam Sterilization, Rectangular Shaped with Steam Indicator, Keystone Cleanroom Products
Covers
Save time and standardize the activities in equipment preparation for sterilization. Leave the covers in place during installation to prevent microbial and particulate contamination. Leave in place while the remaining fill line set-up activities are performed.
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs
  • Individually poly bagged
  • Simplifies the conformance to wrapping SOP's
  • Steam sterilization indicator - autoclavable up to 135 °C
  • Tear and puncture resistant
  • Significant process improvement compared to wrapping with bioshield paper
Elasticized BHD covers provide a convenient, time-saving method of wrapping difficult to wrap product contact change parts prior to steam sterilization.
After sterilization and after the outer bag is removed, leave cover in place during the installation to prevent microbial and particulate contamination. Remove after all other filling line set-up activities are completed. Remaining in place over the equipment parts during setup, their use minimizes microbial or particulate contamination risks.
Color: Blue
Certifications: Certificates of conformance sent with each shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty