Would you like to log in again?
Sharpie® Gamma Irradiated Cleanroom Markers, Keystone Cleanroom Products
Supplier: Keystone Cleanroom Products
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sharpie®
S05UFMARKBLK S01UFMARKBLUE S05FMARKBLK S10UFMARKBLK S02FMARKBLUE S05FMARKBLUE S01FMARKBLK S10FMARKBLK S01RETMARKBLUE S02UFMARKBLK S02UFMARKBLUE S01RETMARKBLK S05UFMARKBLUE S02FMARKBLK S01FMARKBLUE S01UFMARKBLK S10FMARKBLUE S10UFMARKBLUE
76451-892CS 3875.11 USD InStock
76451-892 76451-894 76451-896 76451-920 76451-898 76451-932 76451-910 76451-900 76451-922 76451-912 76451-934 76451-924 76451-902 76451-914 76451-936 76451-926 76451-938 76451-908
Sharpie® Gamma Irradiated Cleanroom Markers, Keystone Cleanroom Products
Pens Cleanroom Pens
Ready to use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek® peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing intruments
Keystone CRP offers gamma irradiated pens and markers for the convenience of pharmaceutical cleanroom operations. These are a 'Ready to Use Option' when sterilization on-site is not possible; or surface sterilization is insufficient. The clean Tyvek® peel pouch packaging allows for effective surface decontamination and easy introduction into the cleanroom through the material handling airlock.
Certifications: Certificates of Irradiation and Conformance with each shipment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty