Would you like to log in again?
VWR CRYO RACK SINGLE UNIT CANISTER 2SQ S
Supplier: VWR International
Cryo rack, single unit canister, Includes 2 square canisters, 50 canes without sleeves, and 32 canes with sleeves, Dimensions: 2.8in x 3 x 24.8in, For high volume storage of vials in canes; maximizing storage capacity in auto-fill tanks, Small
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Price Quantity
VWR CRYO RACK SINGLE UNIT CANISTER 2SQ S 76451-954 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty