Would you like to log in again?
Vials with Anticoagulant Additive for Blood Sample Collection, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HL100 ECV-2.5 ECVK2-0.8-25
76452-022PK 387.3 USD InStock
76452-022 76452-024 76452-020
Vials with Anticoagulant Additive for Blood Sample Collection, SAI Infusion Technologies
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These borosilicate glass vials are a perfect fit for certain automated blood sampling systems. Available with pharmaceutical grade heparin, EDTA, citrate, or your choice of anticoagulant. Formulated for various blood volumes with a variety of additives. Can be customized.
  • Maximize plasma or serum recovery from the smallest blood samples
  • Anticoagulant type and amount are customizable
  • Provided sterile
These extremely small (6×32 mm) borosilicate glass vials provide maximum plasma or serum recovery from the smallest blood samples, from 10 to 300 µl. Anticoagulant type and amount may be customized for your specific study needs.
Packaging: Packed as 100 vials/box.
Caution: Product is for laboratory animal research only.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty