Would you like to log in again?
Diamond®MAX™ Centrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6296 6297 6299 6295 6298
76452-248CS 185.27 USD InStock
76452-248 76452-246 76452-254 76452-252 76452-250
Diamond®MAX™ Centrifuge Tubes, Globe Scientific
Tubes Centrifuge Tubes
Diamond® Max™ Centrifuge tubes are engineered to meet the demands of the most critical scientific work. Premium grade materials and an automated, touch free manufacturing process provide both performance and savings to users.
  • Ultra high performance: High spin rates with maximum RCF ratings of up to 20000 ×g
  • Premium materials: USP Class VI resin is also FDA CFR 21 compliant for food safety
  • Leak resistant: Easy Grip Caps have an integral seal ring design for secure closures that eliminate seepage when used as recommended
  • Value: Efficient automated production reduces cost and assures high quality
  • Eco friendly: Paperboard racks are readily recyclable in the well-established paper recycling stream
The exceptionally clear tubes have black printed graduation marks to full volume of 15 or 50 ml, a bright white marking space, and are packaged in heavy duty, zip, re-closable bags.
Certifications: Tubes are manufactured in a class 100000 clean room, gamma irradiated to SAL 10⁻⁶, and are certified to be free from DNase, RNase, Human DNA, Pyrogens, and Endotoxins.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty