Would you like to log in again?
CAP CARRIER 48 WAY PC BLACK STERILE CS8
Supplier: Greiner Bio-One
Cap Carrier, 48 way, Material: pc, Colour: Black, sterile, for storage of cryo s screw caps
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAP CARRIER 48 WAY PC BLACK STERILE CS8 76452-300 803507 Case of 8 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty